missing translation for 'onlineSavingsMsg'
Learn More

Cilazapril monohydrate, MedChemExpress

Product Code. 16468278
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16468278

Brand: Medchem Express HYA0043A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cilazapril Monohydrate is a angiotensin-converting enzyme (ACE) inhibitor used for the treatment of hypertension and congestive heart failure.

Cilazapril Monohydrate is a angiotensin-converting enzyme (ACE) inhibitor used for the treatment of hypertension and congestive heart failure.

Specifications

Chemical Name or Material Cilazapril monohydrate
Physical Form Powder
CAS 92077-78-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H33N3O6
For Use With (Application) COVID-19-anti-virus
Synonym Ro 31-2848 monohydrate
Solubility Information DMSO : ≥ 100 mg/mL (229.62 mM)
SMILES O=C([C@@H]1CCCN2N1C([C@@H](N[C@H](C(OCC)=O)CCC3=CC=CC=C3)CCC2)=O)O.O
Molecular Weight (g/mol) 435.51
Formula Weight 435.51
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.