missing translation for 'onlineSavingsMsg'
Learn More

CIITA Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£200.00 - £471.00

Specifications

Antigen CIITA
Concentration 1 mg/mL
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Methylated DNA Immunoprecipitation 1:10-1:1000
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18013306
View Documents
Novus Biologicals
NBP2-59072-0.025MG
0.025 mg
£200.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18272465
View Documents
Novus Biologicals
NBP2-59072-0.1MG
0.1 mg
£471.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CIITA Polyclonal specifically detects CIITA in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CIITA
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Methylated DNA Immunoprecipitation 1:10-1:1000
Polyclonal
Rabbit
Human, Mouse, Rat
C2TA, C2TAMHC class II transactivator type III, CIITAIV, class II, major histocompatibility complex, transactivator, MHC class II transactivator, MHC2TANLR family, acid domain containing, NLRA, nucleotide-binding oligomerization domain, leucine rich repeat and acid domaincontaining
CIITA
IgG
Affinity Purified
125 kDa
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
PBS with 0.02% Sodium Azide
4261
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CIITA. The immunogen is located within amino acids 120 - 170 of CIITA.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.