missing translation for 'onlineSavingsMsg'
Learn More

Cig2 Mouse anti-S. pombe, Clone: 3A11/5, Novus Biologicals™

Product Code. p-200084324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18609767 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609767 Supplier Novus Biologicals Supplier No. NBP307584100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cig2 Monoclonal specifically detects Cig2 in S. pombe samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cig2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3A11/5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, ELISA
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Bacterial S. pombe Cig2 Purified protein.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species S. pombe
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.