missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cig2 Monoclonal specifically detects Cig2 in S. pombe samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Cig2 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 3A11/5 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA |
| Formulation | 10 mM PBS with 0.05% BSA |
| Host Species | Mouse |
| Immunogen | Bacterial S. pombe Cig2 Purified protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?