missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cidofovir, MedChemExpress
Description
Cidofovir (GS 0504) is an acyclic monophosphate nucleotide analogue and CMV inhibitor with antiviral activity. Cidofovir inhibits cytomegalovirus (CMV) replication by selectively inhibiting viral DNA polymerase. Cidofovir induces apoptosis and can be used in studies of AIDS cytomegalovirus retinitis, herpes, and cancer. Cidofovir also has anti-orthopoxvirus and anti-variola activities.
Specifications
Specifications
| Chemical Name or Material | Cidofovir |
| Physical Form | Solid |
| CAS | 113852-37-2 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H14N3O6P |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | GS 0504 HPMPC (S)-HPMPC |
| Solubility Information | H2O : 3.33 mg/mL (11.93 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?