missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIDEC Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149276
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
DNA fragmentation is one of the critical steps in apoptosis, which is induced by DNA fragmentation factor (DFF). DFF is composed of two subunits, a 40 kDa caspase-activated nuclease (DFF40) and a 45 kDa inhibitor (DFF45). Recently a novel family of cell-death-inducing DFF45-like effectors (CIDEs) has been identified. Human CIDE-3 is a novel member of CIDEs. There are 2 transcripts, CIDE-3 and CIDE-3alpha, were present in HepG2 and A375 cells. Consistent with its chromosome localization at 3p25, a region associated with high frequency loss of heterozygosity in many tumours, CIDE-3 may play an important role in prevention of tumorigenesis.
Specifications
| CIDEC | |
| Polyclonal | |
| Unconjugated | |
| CIDEC | |
| cell death activator CIDE-3; cell death inducing DFFA like effector c; cell death-inducing DFFA-like effector c; cell death-inducing DFFA-like effector protein C; CID; cide 3; CIDE3; CIDE-3; CIDE-3alpha; Cidec; fat specific gene 27; fat specific protein 27; fat-specific protein FSP27; Fat-specific protein FSP27 homolog; FPLD5; FSP27 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 63924 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96AQ7 | |
| CIDEC | |
| E. coli-derived recombinant human CIDEC, Met75-His176. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction