missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEC Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-430JF646
This item is not returnable.
View return policy
Description
CIDEC Polyclonal antibody specifically detects CIDEC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CIDEC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Janelia Fluor 646 | |
| cell death activator CIDE-3, cell death-inducing DFFA-like effector c, Cell death-inducing DFFA-like effector protein C, CIDE-3, CIDEX, Fat-specific protein FSP27 homolog, FLJ20871, Fsp27, FSP27fat specific protein 27 | |
| Residues 10-25 LLYPKSLSRHVSVRTS] of the 27 kDa human FSP27 (CIDE-3) protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair | |
| 63924 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction