missing translation for 'onlineSavingsMsg'
Learn More

CIDEC Antibody [DyLight 350], Novus Biologicals™

Product Code. 30735189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735189 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735189 Supplier Novus Biologicals Supplier No. AF6960D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CIDEC Polyclonal antibody specifically detects CIDEC in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIDEC
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias cell death activator CIDE-3, cell death-inducing DFFA-like effector c, Cell death-inducing DFFA-like effector protein C, CIDE-3, CIDEX, Fat-specific protein FSP27 homolog, FLJ20871, Fsp27, FSP27fat specific protein 27
Host Species Sheep
Immunogen E. coli-derived recombinant human CIDEC, Met75-His176, Accession # Q96AQ7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 63924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.