missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIDEB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595886
This item is not returnable.
View return policy
Description
Immunogen sequence: DGTAVDSEDF FQLLEDDTCL MVLQSGQSWS PTRSGVLSYG LGRERPKHSK DIARFTFDVY KQNPRDLFGS LNVKATFYGL Y.
CIDEB activates apoptosis.
Specifications
| CIDEB | |
| Polyclonal | |
| Unconjugated | |
| CIDEB | |
| 1110030C18Rik; AI790179; Cell death activator CIDE-B; cell death-inducing DFFA-like effector B; cell death-inducing DNA fragmentation factor, alpha subunit-like effector B; CID; Cideb; CIDE-B; DFFA-like B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27141 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.26 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9UHD4 | |
| CIDEB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-160 of human CIDEB (NP_001305736). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction