missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIDEB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551551
This item is not returnable.
View return policy
Description
Immunogen sequence: MEYLSALNPS DLLRSVSNIS SEFGRRVWTS APPPQRPFRV CDHKRTIRKG LTAATRQELL AKALETLLLN GVLTLVLEED GTAVDSEDFF QLLEDDTCLM VLQSGQSWSP TRSGV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 69%.
CIDEB activates apoptosis.
Specifications
| CIDEB | |
| Polyclonal | |
| Unconjugated | |
| CIDEB | |
| 1110030C18Rik; AI790179; Cell death activator CIDE-B; cell death-inducing DFFA-like effector B; cell death-inducing DNA fragmentation factor, alpha subunit-like effector B; CID; Cideb; CIDE-B; DFFA-like B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27141 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHD4 | |
| CIDEB | |
| Recombinant protein corresponding to Human CIDEB. Recombinant protein control fragment (Product #RP-88872). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction