missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13170-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 25 kDa, and this antibody specifically recognizes the 25 kDa protein.
This gene encodes the homolog of the mouse protein Cidea that has been shown to activate apoptosis. This activation of apoptosis is inhibited by the DNA fragmentation factor DFF45 but not by caspase inhibitors. Mice that lack functional Cidea have higher metabolic rates, higher lipolysis in brown adipose tissue and higher core body temperatures when subjected to cold. These mice are also resistant to diet-induced obesity and diabetes. This suggests that in mice this gene product plays a role in thermogenesis and lipolysis. Alternatively spliced transcripts have been identified.Specifications
| CIDEA | |
| Polyclonal | |
| Unconjugated | |
| CIDEA | |
| Cell death activator CIDE A, CIDE A, CIDEA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1149, 12683, 291541 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60543, O70302 | |
| Cidea | |
| CIDEA Fusion Protein Ag3824 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction