missing translation for 'onlineSavingsMsg'
Learn More

CID5721353, MedChemExpress

Product Code. 16439616
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439616

Brand: Medchem Express HY100502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CID5721353 is an inhibitor of BCL6 with an IC50 value of 212 μM, which corresponds to a Ki of 147 μM.

CID5721353 is an inhibitor of BCL6 with an IC50 value of 212 μM, which corresponds to a Ki of 147 μM.

Specifications

Chemical Name or Material CID5721353
CAS 301356-95-6
Color Brown
Purity Grade Notes Research
Molecular Formula C15H9BrN2O6S2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (109.34 mM; Need ultrasonic)
SMILES O=C(O)C(N(C/1=O)C(SC1=C2C(NC3=C\2C=C(Br)C=C3)=O)=S)CC(O)=O
Molecular Weight (g/mol) 457.28
Formula Weight 457.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.