missing translation for 'onlineSavingsMsg'
Learn More

CID-797718, MedChemExpress

Product Code. 16495038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495038 10 mg 10mg
16461277 50 mg 50mg
16447767 5 mg 5mg
16425786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495038 Supplier Medchem Express Supplier No. HY12243
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CID-797718 is a compound with unknown details.

CID-797718 is a compound with unknown details.

Specifications

Chemical Name or Material CID-797718
CAS 370586-05-3
Color Brown
Purity Grade Notes Research
Molecular Formula C12H11NO3
Quantity 10 mg
Solubility Information DMSO : ≥ 49 mg/mL (225.58 mM)
SMILES O=C1OC2=CC=C(O)C=C2C3=C1NCCC3
Molecular Weight (g/mol) 217.22
Formula Weight 217.22
Percent Purity 99.09%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.