missing translation for 'onlineSavingsMsg'
Learn More

CID 2745687, Tocris Bioscience™

Catalog Number p-4816982
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
264233-05-8
C17H19F2N5O2S
395.429
CYNLZIBKERMMOA-AWQFTUOYSA-N
1-2,4-difluorophenyl-5-2-1,1-dimethylehyl amino thioxomethyl hydrazinylidene methyl-1h-pyrazole-4-carboxylic acid methyl ester, methyl 5-2-tert-butylamino carbothioyl carbohydrazonoyl-1-2,4-difluorophenyl-1h-pyrazole-4-carboxylate, ml hplc, 5-e-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylamino-sulfanylidenemethyl hydrazinylidene methyl-1-2,4-difluorophenyl-4-pyrazolecarboxylic acid methyl ester, methyl 1-2,4-difluoro-phenyl-5-4-tert-butyl-thiosemicarbazono methyl-1h-pyrazole-4-carboxylate, methyl 1-2,4-bis fluoranyl phenyl-5-tert-butylcarbamothioylhydrazinylidene methyl pyrazole-4-carboxylate
9581011
methyl 5-[(E)-(tert-butylcarbamothioylhydrazinylidene)methyl]-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
CC(C)(C)NC(=S)NN=CC1=C(C=NN1C2=C(C=C(C=C2)F)F)C(=O)OC
This item is not returnable. View return policy
264233-05-8
C17H19F2N5O2S
395.429
CYNLZIBKERMMOA-AWQFTUOYSA-N
1-2,4-difluorophenyl-5-2-1,1-dimethylehyl amino thioxomethyl hydrazinylidene methyl-1h-pyrazole-4-carboxylic acid methyl ester, methyl 5-2-tert-butylamino carbothioyl carbohydrazonoyl-1-2,4-difluorophenyl-1h-pyrazole-4-carboxylate, ml hplc, 5-e-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylamino-sulfanylidenemethyl hydrazinylidene methyl-1-2,4-difluorophenyl-4-pyrazolecarboxylic acid methyl ester, methyl 1-2,4-difluoro-phenyl-5-4-tert-butyl-thiosemicarbazono methyl-1h-pyrazole-4-carboxylate, methyl 1-2,4-bis fluoranyl phenyl-5-tert-butylcarbamothioylhydrazinylidene methyl pyrazole-4-carboxylate
9581011
methyl 5-[(E)-(tert-butylcarbamothioylhydrazinylidene)methyl]-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
CC(C)(C)NC(=S)NN=CC1=C(C=NN1C2=C(C=C(C=C2)F)F)C(=O)OC

GPR35 antagonist

CID 2745687 is a competitive, reversible antagonist of the orphan receptor GPR35 (Ki = 12.8 nM). Blocks the GPR35-mediated increase in ERK1/2 phosphorylation and β-arrestin recruitment induced by pamoic acid.
Quantity 10 mg
Formula Weight 395.43
Percent Purity >98%
Chemical Name or Material CID 2745687
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'