missing translation for 'onlineSavingsMsg'
Learn More

CID 2011756, MedChemExpress

Product Code. 16478387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478387 5 mg 5mg
16451857 50 mg 50mg
16435698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478387 Supplier Medchem Express Supplier No. HY13454
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CID 2011756 is an ATP competitive PKD inhibitor, with an IC50 of 3.2 μM for PKD1 in cell free assay, and also shows cellular pan-PKD inhibitory activity against PKD2 and PKD3 (IC50, 0.6 and 0.7 μM, respectively). CID 2011756 also has antitumor activity.

CID 2011756 is an ATP competitive PKD inhibitor, with an IC50 of 3.2 μM for PKD1 in cell free assay, and also shows cellular pan-PKD inhibitory activity against PKD2 and PKD3 (IC50, 0.6 and 0.7 μM, respectively). CID 2011756 also has antitumor activity.

Specifications

Chemical Name or Material CID 2011756
CAS 638156-11-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H21ClN2O3
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (50.39 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C2=CC=CC(Cl)=C2)O1)NC3=CC=C(CN4CCOCC4)C=C3
Molecular Weight (g/mol) 396.87
Formula Weight 396.87
Percent Purity 96.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.