missing translation for 'onlineSavingsMsg'
Learn More

Ciclosporin, Mikromol™

Product Code. 18093970
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093970 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093970 Supplier Mikromol Supplier No. MM0241.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ciclosporin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclosporin A
CAS 59865-13-3
Analyte or Component Names Cyclosporin A
Shipping Condition Room Temperature
Molecular Formula C62 H111 N11 O12
Certifications/Compliance ISO 17025
Synonym Ciclosporin, Cyclo[[(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)oct-6-enoyl]-l-2-aminobutanoyl-N-methylglycyl-N-methyl-l-leucyl-l-valyl-N-methyl-l-leucyl-l-alanyl-d-alanyl-N-methyl-l-leucyl-N-methyl-l-leucyl-N-methyl-l-valyl], ciclosporin A, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., Abrammune, Antibiotic S 7481F1, Arpimune ME, CICPORAL, Cicloral, Cicloral (antibiotic), Ciclosporina Germed, Cipol N, Consupren, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl], Cyclokat, Cyclosporin, Cyclosporine, Cyclosporine A, Debio088, Equoral, Gengraf, Imusporin, NSC 290193, Neoplanta, Neoral, OL 27-400, Optimmune, Papilock Mini, Ramihyphin A, Restasis, S-Neoral, SDZ-OXL 400, Sandimmun, Sandimmun Neoral, Sandimmun Optoral, Sandimmune, Sandimmune Neoral, Sang-35, SangCyA, Sigmasporin Microoral, Zinograf ME
InChI Formula InChI=1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1
SMILES CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-ethyl-33-[(E,1R,2R)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
Molecular Weight (g/mol) 1202.61
Quantity 250 mg
Formula Weight 1201.8414 g/mol
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.