missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciclopirox olamine, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Ciclopirox olamine (Ciclopirox ethanolamine) is a synthetic antifungal agent that can be used for superficial mycoses reseaech. Ciclopirox olamine has a very broad spectrum of activity and inhibits dermatophytes, yeasts, molds, and many Gram-positive and Gram-negative species pathogenic.
Specifications
Specifications
| Chemical Name or Material | Ciclopirox olamine |
| Physical Form | Solid |
| CAS | 41621-49-2 |
| Color | Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H24N2O3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Ciclopirox ethanolamine HOE 296 |
| Solubility Information | DMSO : 25 mg/mL (93.16 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction