missing translation for 'onlineSavingsMsg'
Learn More

Ciclopirox EP Impurity B, Mikromol™

Product Code. 30051062
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051062 25 mg 25mg
17928924 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051062 Supplier Mikromol Supplier No. MM0418.020025
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ciclopirox EP Impurity B
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-cyclohexyl-4-methyl-2H-pyran-2-one
CAS 14818-35-0
Analyte or Component Names 6-cyclohexyl-4-methyl-2H-pyran-2-one
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H16 O2
Certifications/Compliance ISO 9001
Synonym 6-Cyclohexyl-4-methyl-2H-pyran-2-one, Ciclopirox Olamine Imp. B (EP), Ciclopirox USP Related Compound B, Ciclopirox USP RC B, 6-Cyclohexyl-4-methyl-2H-pyran-2-one, Ciclopirox Related Compound B (USP), Ciclopirox impurity B (EP), 6-Cyclohexyl-4-methyl-2-pyrone, Ciclopirox Related Compound B, Ciclopirox Olamine Impurity B, Ciclopirox Impurity B
InChI Formula InChI=1S/C12H16O2/c1-9-7-11(14-12(13)8-9)10-5-3-2-4-6-10/h7-8,10H,2-6H2,1H3
SMILES CC1=CC(=O)OC(=C1)C2CCCCC2
IUPAC Name 6-cyclohexyl-4-methylpyran-2-one
Molecular Weight (g/mol) 192.25
Quantity 25 mg
Formula Weight 192.115 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.