missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIB1, Mouse anti-Human, Clone: 1D1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB1113.100uL
This item is not returnable.
View return policy
Specifications
| CIB1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_006384 | |
| CIB1 | |
| Recombinant protein corresponding to full length human CIB1. | |
| RUO | |
| 10519 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 1D1 | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| calcium and integrin binding 1 (calmyrin) | |
| CIB/KIP/KIP1/SIP2-28 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction