missing translation for 'onlineSavingsMsg'
Learn More

CIB1 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30731580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731580 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731580 Supplier Novus Biologicals Supplier No. AF7557E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CIB1 Polyclonal antibody specifically detects CIB1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIB1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28
Host Species Sheep
Immunogen E. coli-derived recombinant human CIB1, Gly2-Leu191, Accession # Q99828
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10519
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.