missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIB1 Antibody (1D1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04279AF532
This item is not returnable.
View return policy
Description
CIB1 Monoclonal specifically detects CIB1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CIB1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CIB1 | |
| Recombinant human CIB1 (1-191aa) purified from E. coli | |
| Signal Transduction | |
| 10519 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1D1 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction