missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIAPIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559903
This item is not returnable.
View return policy
Description
Immunogen sequence: KLWTLSANDM EDDSMDLIDS DELLDPEDLK KPDPASLRAA SCGEGKKRKA CKNCTCGLAE ELEKEKSREQ MSSQPKSACG NCYLGDAFRC ASCPYLGMPA FKPGEKVLLS D Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.
Specifications
| CIAPIN1 | |
| Polyclonal | |
| Unconjugated | |
| CIAPIN1 | |
| 2810413N20Rik; AA617265; Anamorsin; anamorsin {ECO:0000255; AU021794; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; BHLHB2; Bhlhe40; C130042M06Rik; CIAPIN1; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; CUA001; cytokine induced apoptosis inhibitor 1; cytokine response gene 8; cytokine-induced apoptosis inhibitor 1; cytokine-induced apoptosis inhibitor 1 {ECO:0000255; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; DRE2; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of-split and hairy-related protein 2; Fe-S cluster assembly protein DRE2 homolog; fe-S cluster assembly protein DRE2 homolog {ECO:0000255; HAMAP-Rule:MF_03115}; HLHB2; predicted protein of HQ0915; PRO0915; Sharp2; SHARP-2; Stimulated by retinoic acid gene 13 protein; Stra13; Stra14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109006, 307649, 57019 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XID1, Q6FI81, Q8WTY4 | |
| CIAPIN1 | |
| Recombinant protein corresponding to Human Anamorsin. Recombinant protein control fragment (Product #RP-98549). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction