missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ cIAP1 Recombinant Rabbit Monoclonal Antibody (K01_1P56)

Product Code. 30830269
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830269 50 μL
30827896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830269 Supplier Invitrogen™ Supplier No. MA565323
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

cIAP1 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1P56
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene BIRC2
Gene Accession No. Q13490
Gene Alias Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW146227; baculoviral IAP repeat containing 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 2; baculoviral IAP repeat-containing protein 2; putative inhibitor of apoptosis; BIRC2; Birc3; cellular inhibitor of apoptosis 1; cIAP1; C-IAP1; cIAP2; HIAP1; HIAP2; Hiap-2; IAP homolog B; IAP1; IAP2; IAP-2; inhibitor of apoptosis protein; inhibitor of apoptosis protein 2; LOC100622859; mcIAP1; MIAP1; MIAP2; mIAP-2; MIHB; MIHC; NFR2-TRAF signalling complex protein; rIAP1; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; RNF48; TNFR2-TRAF-signaling complex protein 2
Gene Symbols BIRC2
Host Species Rabbit
Immunogen A synthetic peptide of human cIAP1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.