missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP-2/HIAP-1 Antibody (SR2173), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21634-50ul
This item is not returnable.
View return policy
Description
cIAP-2/HIAP-1 Monoclonal antibody specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| cIAP-2/HIAP-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR2173 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:50 | |
| AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49 | |
| A synthesized peptide derived from human cIAP-2/HIAP-1 (Uniprot #: Q13489) | |
| 50 μg | |
| Apoptosis, Cancer, Inhibitors Activators and Regulators, Zinc Finger | |
| 330 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction