missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP-2/HIAP-1 Antibody (OTI1A9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70184CL1
This item is not returnable.
View return policy
Description
cIAP-2/HIAP-1 Monoclonal antibody specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| cIAP-2/HIAP-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49 | |
| Human recombinant protein fragment corresponding to amino acids 255-529 of human BIRC3 (NP_001156) produced in E.coli. | |
| 0.1 mL | |
| Apoptosis, Cancer, Inhibitors Activators and Regulators, Zinc Finger | |
| 330 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI1A9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction