missing translation for 'onlineSavingsMsg'
Learn More

cIAP-1/HIAP-2 Rabbit anti-Human, Rat, Clone: 2L0W9, Novus Biologicals™

Product Code. 18379344 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379344 20 μg 20µL
18360472 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379344 Supplier Novus Biologicals Supplier No. NBP31586020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

cIAP-1/HIAP-2 Monoclonal antibody specifically detects cIAP-1/HIAP-2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP-1/HIAP-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2L0W9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human cIAP-1/HIAP-2 (Q13490).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 329
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.