missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP-1/HIAP-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77193
This item is not returnable.
View return policy
Description
cIAP-1/HIAP-2 Polyclonal specifically detects cIAP-1/HIAP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| cIAP-1/HIAP-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2 | |
| Rabbit | |
| 70 kDa | |
| 0.1 mg | |
| Apoptosis | |
| 329 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-4 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin | |
| Q13490 | |
| BIRC2 | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of human c-IAP1. c-IAP antibody detects both c-IAP1 and c-IAP2. The immunogen is located within amino acids 540 - 590 of cIAP. Amino Acid Squence: SLEEQLRRLQEERT | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction