missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CIAO1 Polyclonal specifically detects CIAO1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | CIAO1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | CIA1, cytosolic iron-sulfur protein assembly 1, cytosolic iron-sulfur protein assembly 1 homolog (S. cerevisiae), probable cytosolic iron-sulfur protein assembly protein CIAO1, WD repeat domain 39, WD repeat-containing protein 39, WD40 protein Ciao1, WDR39cytosolic iron-sulfur protein assembly 1 homolog |
| Gene Symbols | CIAO1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:CSDDRTVRIWRQYLPGNEQGVACSGSDPSWKCICTLSGFHSRTIYDIAWCQLTGALATACGDDAIRVFQEDPNSDPQQPTFSLTAHLHQAHSQDVNCVAWNPKEPGLLASCSDDGEVAFWKYQR |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?