missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CI016 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570935
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 83%.
C9orf16 is a protein coding gene.
Specifications
| CI016 | |
| Polyclonal | |
| Unconjugated | |
| C9orf16 | |
| C9orf16; chromosome 9 open reading frame 16; EST00098; UPF0184 protein C9orf16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79095 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BUW7 | |
| C9orf16 | |
| synthetic peptide directed towards the middle region of human CI016. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu