missing translation for 'onlineSavingsMsg'
Learn More

CI 994, Thermo Scientific™

Product Code. 15416969
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416969 100 mg 100mg
15406969 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416969 Supplier Thermo Scientific Alfa Aesar Supplier No. J65561.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112522-64-2
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00866266
InChI Key VAZAPHZUAVEOMC-UHFFFAOYSA-N
Synonym tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
PubChem CID 2746
IUPAC Name 4-acetamido-N-(2-aminophenyl)benzamide
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N

Specifications

Color White
Quantity 100 mg
Sensitivity Air sensitive
Solubility Information Soluble in DMSO
Formula Weight 269.3
Physical Form Powder
Chemical Name or Material CI 994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.