missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CI 966 hydrochloride

Description
Selective inhibitor of the GABA transporter GAT-1 (IC50 values are 0.26 and 1.2 μM at cloned human and rat GAT-1 respectively). Displays over 200-fold selectivity over GAT-2 and GAT-3. Centrally active upon systemic administration in vivo. Anticonvulsive and neuroprotective.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 509.87 |
| Purity | >98% |
| Molecular Weight (g/mol) | 509.873 |
| Chemical Name or Material | CI 966 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?