missing translation for 'onlineSavingsMsg'
Learn More

CI-1044, MedChemExpress

Product Code. 16445617
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16445617

Brand: Medchem Express HY100246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CI-1044 is an orally active PDE4 inhibitor with IC50s of 0.29, 0.08, 0.56, 0.09 μM for PDE4A5, PDE4B2, PDE4C2 and PDE4D3, respectively.

CI-1044 is an orally active PDE4 inhibitor with IC50s of 0.29, 0.08, 0.56, 0.09 μM for PDE4A5, PDE4B2, PDE4C2 and PDE4D3, respectively.

Specifications

Chemical Name or Material CI-1044
CAS 197894-84-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H19N5O2
Quantity 5 mg
Synonym PD-189659
Solubility Information DMSO : 125 mg/mL (314.52 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)N[C@H]2C(N3C4=C(CC3)C=C(N)C=C4C(C5=CC=CC=C5)=N2)=O
Molecular Weight (g/mol) 397.43
Formula Weight 397.43
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.