missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymotrypsin-like protease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | Chymotrypsin-like protease |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18466111
|
Novus Biologicals
NBP2-33973-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105673
|
Novus Biologicals
NBP2-33973 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chymotrypsin-like protease Polyclonal specifically detects Chymotrypsin-like protease in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Chymotrypsin-like protease | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| chymotrypsin-like, chymotrypsin-like protease CTRL-1, CTRL1, EC 3.4.21, EC 3.4.21.-, MGC70821 | |
| CTRL | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P40313 | |
| 1506 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VTAAHCNVSPGRHFVVLGEYDRSSNAEPLQVLSVSRAITHPSWNSTTMNNDVTLLK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title