missing translation for 'onlineSavingsMsg'
Learn More

Chymotrypsin C/CTRC Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18320142 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320142 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320142 Supplier Novus Biologicals Supplier No. NBP299799JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Chymotrypsin C/CTRC Polyclonal specifically detects Chymotrypsin C/CTRC in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chymotrypsin C/CTRC
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Caldecrin, chymotrypsin C (caldecrin), chymotrypsin-C, CLCRserum calcium decreasing factor, EC 3.4.21, EC 3.4.21.2, ELA4, elastase 4, elastase IV
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Chymotrypsin C/CTRC (Q99895; Met1-Leu268).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11330
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.