missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Chymotrypsin Alpha (Bovine)

Product Code. 16141760
Change view
Click to view available options
:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16141760 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16141760 Supplier Abnova™ Supplier No. P5212.100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00766
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 25.6kDa
Name Chymotrypsin Alpha (Bovine)
Preparation Method Native protein purified from Bovine Pancreas according to the procedure of Yapel et al., J. Amer. Chem. Soc., 88, 2573 (1966)
Quantity 100 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name CHYMOTRYPSIN
Assays Chymotrypsin Assay; Sequence analysis; Peptide synthesis; Peptide mapping; Peptide fingerprinting.
Biological Activity ≥45 units per mg protein. One Unit hydrolyzes one micromole of benzoyl-L-tyrosine ethyl ester per minute at 25°C, pH 7.8 in the presence of calcium. An activity of 45 units per mg using the above definition, is the equivalent of 10,000 optical density or 1330 N.F. units per mg using ATEE as a substrate.
1 BTEE unit = 29.5 USP/NF units.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas according to the procedure of Yapel et al., J. Amer. Chem. Soc., 88, 2573 (1966)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.