missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymase/CMA1/Mast Cell Chymase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-27551
336.60 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Chymase/CMA1/Mast Cell Chymase Polyclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Chymase/CMA1/Mast Cell Chymase | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1 ug/mL, Immunohistochemistry 1.5 ug/mL, Immunohistochemistry-Paraffin 1.5 ug/mL, Peptide ELISA Detection limit 1:128000 | |
| NP_001827.1 | |
| CMA1 | |
| Peptide with sequence C-QFRHPKYNTSTLHHD corresponding to internal region according to NP_001827.1. | |
| 0.1 mg | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Mast Cell Markers | |
| 1215 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction