missing translation for 'onlineSavingsMsg'
Learn More

ChX710, MedChemExpress

Product Code. 16495556
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495556 100 mg 100mg
16484738 10 mg 10mg
16473226 25 mg 25mg
16447457 5 mg 5mg
16431027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495556 Supplier Medchem Express Supplier No. HY112951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3.

ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3.

Specifications

Chemical Name or Material ChX710
Physical Form Solid
CAS 2438721-44-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H21N5O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (61.84 mM; Need ultrasonic)
SMILES O=C(NC(C)CN(C)C)C1=C(N=C(C2=CC=NC=C2)N3)C3=CC=C1
Molecular Weight (g/mol) 323.39
Formula Weight 323.39
Percent Purity 99.12%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.