missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHX10 Sheep anti-Human, Mouse, Rat, Chicken, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
£199.00 - £367.00
Specifications
| Antigen | CHX10 |
|---|---|
| Dilution | Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18322346
|
Novus Biologicals
NBP3-11891-25UG |
25 μg |
£199.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18348203
|
Bio-Techne
NBP3-11891-250UG |
250 μg |
£367.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHX10 Polyclonal antibody specifically detects CHX10 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
| CHX10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Sheep | |
| Vision | |
| PBS | |
| 338917 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Chicken | |
| ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2 | |
| Recombinant protein corresponding to amino acids 264 to 361 derived from the C-terminal region of the human CHX10 protein conjugated to KLH. (Uniprot: P58304) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title