missing translation for 'onlineSavingsMsg'
Learn More

CHX10 Sheep anti-Human, Mouse, Rat, Chicken, Polyclonal, Novus Biologicals™

Product Code. 18366382 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
250 μg
Unit Size:
250µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366382 250 μg 250µg
18327423 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18366382 Supplier Bio-Techne Supplier No. NBP311892250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CHX10 Polyclonal antibody specifically detects CHX10 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2
Host Species Sheep
Immunogen Antibody raised in sheep against a recombinant protein corresponding to amino acids 1 to 131 derived from the N-terminal region of the human CHX10 protein conjugated to KLH. (Uniprot: P58304)
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 338917
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.