missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHX10 Sheep anti-Human, Mouse, Rat, Chicken, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CHX10 Polyclonal antibody specifically detects CHX10 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | CHX10 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen |
| Formulation | PBS |
| Gene Alias | ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2 |
| Host Species | Sheep |
| Immunogen | Recombinant protein corresponding to amino acids 264 to 361 derived from the C-terminal region of the human CHX10 protein conjugated to KLH. (Uniprot: P58304) |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?