missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHX10 Recombinant Rabbit Monoclonal Antibody (4Z4R2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538025
This item is not returnable.
View return policy
Description
Positive Samples: Mouse spinal cord, Mouse brain, Rat spinal cord, Rat brain Immunogen sequence: AAAESGRKPE GERQALPKLD KMEQDERGPD AQAAISQEEL RENSIAVLRA KAQEHSTKVL GTVSGPDSLA RSTEKPEEEE AMDEDRPAER LSPPQLEDMA.
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
Specifications
| CHX10 | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide, pH 7.3 | |
| Q61412 | |
| VSX2 | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4Z4R2 | |
| Unconjugated | |
| VSX2 | |
| C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12677, 171360 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction