missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHX10 Recombinant Rabbit Monoclonal Antibody (24GB2590)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557315
This item is not returnable.
View return policy
Description
CHX10 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
Specifications
| CHX10 | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P58304, Q61412 | |
| VSX2 | |
| A synthesized peptide derived from human CHX10 (aa 330-361). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2590 | |
| Unconjugated | |
| VSX2 | |
| C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12677, 171360, 338917 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction