missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHX10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582092
This item is not returnable.
View return policy
Description
Immunogen sequence: LGLNKEPPSS HPRAALDGLA PGHLLAARSV LSPAGVGGMG LLGPGGLPGF YTQPTFLEVL SDPQSVHLQP LGRASGPLDT SQTASSDSED VSSSDRKMSK SALNQTKKRK KRRHRTIFTS YQLEELEKAF NEAH Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
Specifications
| CHX10 | |
| Polyclonal | |
| Unconjugated | |
| VSX2 | |
| C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171360, 338917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P58304 | |
| VSX2 | |
| Recombinant protein corresponding to Human VSX2. Recombinant protein control fragment (Product #RP-89128). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction