missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHX10 Antibody (SR1872), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | CHX10 |
|---|---|
| Clone | SR1872 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673215
|
Novus Biologicals
NBP3-22295-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602454
|
Novus Biologicals
NBP3-22295-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHX10 Monoclonal antibody specifically detects CHX10 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| CHX10 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2 | |
| A synthesized peptide derived from human CHX10 (Uniprot #: P58304) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1872 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 338917 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title