missing translation for 'onlineSavingsMsg'
Learn More

CHX10 Antibody, Novus Biologicals™

Product Code. 18416610 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416610 25 μL 25µL
18755743 - 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18416610 Supplier Novus Biologicals Supplier No. NBP18447625ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

CHX10 Polyclonal specifically detects CHX10 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHX10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID 28440470)., Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P58304
Gene Alias ceh-10 homeo domain containing homolog, ceh-10 homeo domain containing homolog (C. elegans), ceh-10 homeodomain containing homolog (C. elegans), Ceh-10 homeodomain-containing homolog, CHX10MCOPCB3, Homeobox protein CHX10, HOX10C elegans ceh-10 homeo domain-containing homolog, MCOP2, RET1, visual system homeobox 2
Gene Symbols VSX2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LGLNKEPPSSHPRAALDGLAPGHLLAARSVLSPAGVGGMGLLGPGGLPGFYTQPTFLEVLSDPQSVHLQPLGRASGPLDTSQTASSDSEDVSSSDRKMSKSALNQTKKRKKRRHRTIFTSYQLEELEKAFNEAH
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 338917
Test Specificity Specificity of human CHX10 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.