missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHURC1, Mouse anti-Human, Clone: 2F9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00091612-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MCGDCVGKEYPNRGNTCLENGSFLLNFTGCAVCSKRDFMLITNKSLKEEDGEEIVTYDHLCKNCHHVIARHEYTFSIMDEFQEYTMLCLLCGKAEDTISILPDDPRQMTLLFSpecifications
| CHURC1 | |
| Monoclonal | |
| Unconjugated | |
| churchill domain containing 1 | |
| C14orf52/FLJ33064/My015/chch | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2F9 | |
| In 1x PBS, pH 7.4 | |
| BC020550 | |
| CHURC1 | |
| CHURC1 (AAH20550, 1 a.a. ∼ 112 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 91612 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction