Learn More
Caframo™ Chuck Guard
Description
- Magnets securely snap guard into place on stirrer housing over the chuck
- Creates a barrier around the spinning chuck
- Chemical-resistant polyurethane
- Easy to install and remove
- Protects users from the moving chuck
- Protects the chuck from splashes
Specifications
Specifications
| For Use With (Equipment) | Use with overhead stirrers BDC1850, BDC3030, and BDC6015. |
| For Use With (Application) | Creates barrier around moving chuck, as well as protects chuck from chemical splashes. |
| Model | CG1 |
| Quantity | Each |
| Type | Chuck Guard |
| Weight (Metric) | 14 g |
| Material | Plastic |