missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHTF8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CHTF8 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274323
|
Novus Biologicals
NBP2-56494 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671558
|
Novus Biologicals
NBP2-56494-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHTF8 Polyclonal specifically detects CHTF8 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CHTF8 | |
| Polyclonal | |
| Rabbit | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 54921 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MGPGSGPNLRAGVLLTSGNGPPNPRPVGLGPGPNPNLRSGFLGTNPAPRSGVFPGPG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| chromosome transmission fidelity protein 8 homolog, CTF8, chromosome transmission fidelity factor 8 homolog (S. cerevisiae), CTF8hCTF8, Decreased expression in renal and prostate cancer protein, DERPCchromosome transmission fidelity factor 8 homolog, FLJ20400 | |
| CHTF8 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title