missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST7 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547699
This item is not returnable.
View return policy
Description
In direct ELISA, less than 1% cross-reactivity with recombinant mouse (rm) CHST1, recombinant human CHST2, rmCHST5, and rmCHST3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene belongs to the sulfotransferase gene family. Sulfotransferases generate sulfated glycosaminoglycan (GAG) moities during chondroitin sulfate biosynthesis. They create considerable structural diversity among chondroitin sulfates by transferring sulfate with remarkable specificity for the underlying oligosaccharide substrate. This gene product mainly transfers sulfate to N-acetylgalactosamine. The regulated expression of each member of this gene family may be an important determinant of sulfated GAGs expression and the associated function of chondroitin sulfates as regulators of many biologic processes. This gene is part of a gene cluster on chromosome Xp11.23.
Specifications
| CHST7 | |
| Polyclonal | |
| Unconjugated | |
| CHST7 | |
| 2600013M07Rik; C6ST-2; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7; carbohydrate (N-acetylglucosamino) sulfotransferase 7; carbohydrate sulfotransferase 7; Chondroitin 6-sulfotransferase 2; chondroitin 6-sulfotransferase 7; chondroitin 6-sulfotransferase-2; CHST7; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 5; glcNAc6ST-4; Gn6st-4; Gst5; GST-5; mC6ST-2; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine 6-O-sulfotransferase 4 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 60322 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9EP78 | |
| CHST7 | |
| CHO-derived recombinant mouse CHST7 Ser34-Val484. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction