missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18242-1-AP-150UL
This item is not returnable.
View return policy
Description
CHST3 is an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrixand most cells which is involved in cell migration and differentiation.Specifications
| CHST3 | |
| Polyclonal | |
| Unconjugated | |
| CHST3 | |
| C6ST, C6ST 1, C6ST1, Chondroitin 6 sulfotransferase, CHST3, GST 0, HSD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 53374, 84468, 9469 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88199, Q7LGC8, Q9QZL2 | |
| Chst3 | |
| CHST3 Fusion Protein Ag13008 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction